4-Brom-2-(trifluormethyl)benzonitril
Lieferant: COMBI-BLOCKS
OS-7564-1G
OS-7564-5G
COBBOS-7564-1GEA
30.7
EUR
COBBOS-7564-1G
COBBOS-7564-5G
4-Brom-2-(trifluormethyl)benzonitril
4-Brom-2-(trifluormethyl)benzonitril
Formel:
C₈H₃BrF₃N |
MDL:
MFCD06659466 CAS-Nummer: 191165-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...