4-Brom-2-(trifluormethyl)benzonitril

Lieferant: COMBI-BLOCKS

OS-7564-1G OS-7564-5G
COBBOS-7564-1GEA 30.7 EUR
COBBOS-7564-1G COBBOS-7564-5G
4-Brom-2-(trifluormethyl)benzonitril
4-Brom-2-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
MDL: MFCD06659466
CAS-Nummer: 191165-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR