1-Brom-2,4-bis(trifluormethyl)benzol
Lieferant: COMBI-BLOCKS
Synonyme:
2,4-Bis(trifluoromethyl)-1-bromobenzene
OS-7565-5G
OS-7565-25G
OS-7565-100G
COBBOS-7565-5GEA
0
EUR
COBBOS-7565-5G
COBBOS-7565-25G
COBBOS-7565-100G
1-Brom-2,4-bis(trifluormethyl)benzol
1-Brom-2,4-bis(trifluormethyl)benzol
Formel:
C₈H₃BrF₆ |
MDL:
MFCD00074904 CAS-Nummer: 327-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...