1-Brom-2,4-bis(trifluormethyl)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 2,4-Bis(trifluoromethyl)-1-bromobenzene

OS-7565-5G OS-7565-25G OS-7565-100G
COBBOS-7565-5GEA 0 EUR
COBBOS-7565-5G COBBOS-7565-25G COBBOS-7565-100G
1-Brom-2,4-bis(trifluormethyl)benzol
1-Brom-2,4-bis(trifluormethyl)benzol
Formel: C₈H₃BrF₆
MDL: MFCD00074904
CAS-Nummer: 327-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR