2-(Trimethylsilyl)ethoxymethylchlorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Chloromethoxy)ethyltrimethylsilane, 2-(Trimethylsilyl)ethoxymethyl Chloride, SEM-chloride

OS-7574-25G OS-7574-1G OS-7574-5G
COBBOS-7574-25GEA 281 EUR
COBBOS-7574-25G COBBOS-7574-1G COBBOS-7574-5G
2-(Trimethylsilyl)ethoxymethylchlorid
2-(Trimethylsilyl)ethoxymethylchlorid
Formel: C₆H₁₅ClOSi
MDL: MFCD00009919
CAS-Nummer: 76513-69-4
EINECS: 278-483-4
UN: 2920
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR