2-Chlor-5-fluorpyridin

Lieferant: COMBI-BLOCKS

OS-7583-5G OS-7583-25G OS-7583-1G
COBBOS-7583-5GEA 48.8 EUR
COBBOS-7583-5G COBBOS-7583-25G COBBOS-7583-1G
2-Chlor-5-fluorpyridin
Formel: C₅H₃ClFN
MDL: MFCD03095332
CAS-Nummer: 31301-51-6
UN: 2929
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR