1-Brom-4-pentylbenzol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-pentylbenzol, (4-Bromphenyl)pentan, p-Brom-pentylbenzol
OS-7594-5G
OS-7594-25G
OS-7594-100G
COBBOS-7594-5GEA
26.2
EUR
COBBOS-7594-5G
COBBOS-7594-25G
COBBOS-7594-100G
1-Brom-4-pentylbenzol
1-Brom-4-pentylbenzol
Formel:
C₁₁H₁₅Br Molecular Weight: 227.14 g/mol |
MDL:
MFCD00061113 CAS-Nummer: 51554-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...