6-Methylnicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methyl-3-pyridincarbonsäure, 6-Methylpyridin-3-carbonsäure

OS-7595-25G OS-7595-5G
COBBOS-7595-25GEA 0 EUR
COBBOS-7595-25G COBBOS-7595-5G
6-Methylnicotinsäure
6-Methylnicotinsäure
Formel: C₇H₇NO₂
Schmelzpunkt: 212 °C
MDL: MFCD00006341
CAS-Nummer: 3222-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR