1-Brom-4-hexylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: p-Brom-hexylbenzol

OS-7603-5G OS-7603-25G OS-7603-1G
COBBOS-7603-5GEA 60.7 EUR
COBBOS-7603-5G COBBOS-7603-25G COBBOS-7603-1G
1-Brom-4-hexylbenzol
1-Brom-4-hexylbenzol
Formel: C₁₂H₁₇Br
Molecular Weight: 241.17 g/mol
MDL: MFCD00061114
CAS-Nummer: 23703-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR