5-Brom-2-chlorphenol
Lieferant: COMBI-BLOCKS
OS-7612-25G
OS-7612-5G
OS-7612-1G
OS-7612-100G
COBBOS-7612-25GEA
0
EUR
COBBOS-7612-25G
COBBOS-7612-5G
COBBOS-7612-1G
COBBOS-7612-100G
5-Brom-2-chlorphenol
5-Brom-2-chlorphenol
Formel:
C₆H₄BrClO |
MDL:
MFCD00672940 CAS-Nummer: 183802-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...