5-Brom-2-chlorphenol

Lieferant: COMBI-BLOCKS

OS-7612-25G OS-7612-5G OS-7612-1G OS-7612-100G
COBBOS-7612-25GEA 0 EUR
COBBOS-7612-25G COBBOS-7612-5G COBBOS-7612-1G COBBOS-7612-100G
5-Brom-2-chlorphenol
5-Brom-2-chlorphenol
Formel: C₆H₄BrClO
MDL: MFCD00672940
CAS-Nummer: 183802-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR