2,3-Difluoranilin

Lieferant: COMBI-BLOCKS

OS-7615-1G OS-7615-100G OS-7615-25G OS-7615-5G
COBBOS-7615-1GEA 25.5 EUR
COBBOS-7615-1G COBBOS-7615-100G COBBOS-7615-25G COBBOS-7615-5G
2,3-Difluoranilin
2,3-Difluoranilin
Formel: C₆H₅F₂N
Molecular Weight: 129.11 g/mol
Siedepunkt: 69 °C (12 mmHg)
Dichte: 1.274 g/cm³ (20 °C)
MDL: MFCD00010298
CAS-Nummer: 4519-40-8
EINECS: 224-847-2
UN: 2941
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR