2-Brom-3-fluorpyridin

Lieferant: COMBI-BLOCKS

OS-7616-5G OS-7616-25G OS-7616-1G
COBBOS-7616-5GEA 0 EUR
COBBOS-7616-5G COBBOS-7616-25G COBBOS-7616-1G
2-Brom-3-fluorpyridin
2-Bromo-3-fluoropyridine
Formel: C₅H₃BrFN
MDL: MFCD04114128
CAS-Nummer: 40273-45-8
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR