3,5-Difluorpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Difluoropyridine-2-carboxylic acid

OS-7627-5G OS-7627-1G
COBBOS-7627-5GEA 112 EUR
COBBOS-7627-5G COBBOS-7627-1G
3,5-Difluorpicolinsäure
3,5-Difluorpicolinsäure
Formel: C₆H₃F₂NO₂
Molecular Weight: 159.09 g/mol
MDL: MFCD05664124
CAS-Nummer: 745784-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR