3-Brom-2-pyridincarbonitril
Lieferant: COMBI-BLOCKS
Synonyme:
3-Brom-2-pyridincarbonsäurenitril, 3-Brom-2-cyanpyridin
OS-7656-1G
OS-7656-25G
OS-7656-5G
COBBOS-7656-1GEA
33.2
EUR
COBBOS-7656-1G
COBBOS-7656-25G
COBBOS-7656-5G
3-Brom-2-pyridincarbonitril
3-Brom-2-pyridincarbonitril
Formel:
C₆H₃BrN₂ Molecular Weight: 183.01 g/mol |
MDL:
MFCD02683288 CAS-Nummer: 55758-02-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...