1-Chlor-4-(trifluormethoxy)benzol
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Trifluoromethoxy)chlorobenzene
OS-7658-5G
OS-7658-25G
OS-7658-100G
COBBOS-7658-5GEA
0
EUR
COBBOS-7658-5G
COBBOS-7658-25G
COBBOS-7658-100G
1-Chlor-4-(trifluormethoxy)benzol
1-Chloro-4-(trifluoromethoxy)benzene
Formel:
C₇H₄ClF₃O |
MDL:
MFCD00040953 CAS-Nummer: 461-81-4 EINECS: 207-315-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...