1-Chlor-4-(trifluormethoxy)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Trifluoromethoxy)chlorobenzene

OS-7658-5G OS-7658-25G OS-7658-100G
COBBOS-7658-5GEA 0 EUR
COBBOS-7658-5G COBBOS-7658-25G COBBOS-7658-100G
1-Chlor-4-(trifluormethoxy)benzol
1-Chloro-4-(trifluoromethoxy)benzene
Formel: C₇H₄ClF₃O
MDL: MFCD00040953
CAS-Nummer: 461-81-4
EINECS: 207-315-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR