3-(Trifluormethoxy)anisol

Lieferant: COMBI-BLOCKS
Achtung

OS-7664-1G OS-7664-25G OS-7664-5G
COBBOS-7664-1GEA 0 EUR
COBBOS-7664-1G COBBOS-7664-25G COBBOS-7664-5G
3-(Trifluormethoxy)anisol
3-(Trifluormethoxy)anisol
Formel: C₈H₇F₃O₂
MDL: MFCD01631504
CAS-Nummer: 142738-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR