4-Fluor-2-iodanilin

Lieferant: COMBI-BLOCKS

OS-7693-5G OS-7693-25G OS-7693-100G
COBBOS-7693-5GEA 25.9 EUR
COBBOS-7693-5G COBBOS-7693-25G COBBOS-7693-100G
4-Fluor-2-iodanilin
4-Fluor-2-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237.02 g/mol
MDL: MFCD00042584
CAS-Nummer: 61272-76-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR