3-Cyan-4-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methyl-3-pyridincarbonitril, 3-Cyan-4-picolin

OS-7729-100G
COBBOS-7729-100GEA 612 EUR
COBBOS-7729-100G
3-Cyan-4-methylpyridin
3-Cyano-4-methylpyridine
Formel: C₇H₆N₂
Molecular Weight: 118,14 g/mol
MDL: MFCD00234272
CAS-Nummer: 5444-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR