2,6-Dichlorphenylhydraziniumchlorid

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dichlorphenylhydrazinhydrochlorid, 2,6-Dichlorphenylhydrazin Hydrochlorid, 1-(2,6-Dichlorphenyl)hydrazin Hydrochlorid

OS-7730-5G
COBBOS-7730-5GEA 25.5 EUR
COBBOS-7730-5G
2,6-Dichlorphenylhydraziniumchlorid
2,6-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213,49 g/mol
MDL: MFCD00012930
CAS-Nummer: 50709-36-9
EINECS: 256-730-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR