1-Brom-5-chlor-2-iodbenzol

Lieferant: COMBI-BLOCKS

OS-7731-100G
COBBOS-7731-100GEA 336 EUR
COBBOS-7731-100G
1-Brom-5-chlor-2-iodbenzol
1-Brom-5-chlor-2-iodbenzol
Formel: C₆H₃BrClI
Molecular Weight: 317,35 g/mol
MDL: MFCD00672944
CAS-Nummer: 31928-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR