2-Fluor-5-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluor-3-methoxynitrobenzol

OS-7736-5G
COBBOS-7736-5GEA 61 EUR
COBBOS-7736-5G
2-Fluor-5-nitroanisol
2-Fluor-5-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
MDL: MFCD07782081
CAS-Nummer: 454-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR