2-Chlor-4-fluorbenzotrifluorid

Lieferant: COMBI-BLOCKS

OS-7749-1G
COBBOS-7749-1GEA 0 EUR
COBBOS-7749-1G
2-Chlor-4-fluorbenzotrifluorid
2-Chlor-4-fluorbenzotrifluorid
Formel: C₇H₃ClF₄
MDL: MFCD03412197
CAS-Nummer: 94444-58-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR