3-Fluor-4-nitroanilin

Lieferant: COMBI-BLOCKS

OS-7802-5G
COBBOS-7802-5GEA 71.8 EUR
COBBOS-7802-5G
3-Fluor-4-nitroanilin
3-Fluor-4-nitroanilin
Formel: C₆H₅FN₂O₂
Molecular Weight: 156,12 g/mol
MDL: MFCD03412239
CAS-Nummer: 2369-13-3
EINECS: 219-130-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR