2-Brom-5-fluoranisol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 1-Bromo-4-fluoro-2-methoxybenzene

OS-7820-25G
COBBOS-7820-25GEA 130 EUR
COBBOS-7820-25G
2-Brom-5-fluoranisol
2-Brom-5-fluoranisol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD04973752
CAS-Nummer: 450-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR