4-Amino-2-chlorpyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlorpyrimidin-4-ylamin

OS-7833-25G
COBBOS-7833-25GEA 122 EUR
COBBOS-7833-25G
4-Amino-2-chlorpyrimidin
4-Amino-2-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
MDL: MFCD00194051
CAS-Nummer: 7461-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR