4-Amino-3,6-dichlorpyridazin

Lieferant: COMBI-BLOCKS

OS-7882-5G
COBBOS-7882-5GEA 79.1 EUR
COBBOS-7882-5G
4-Amino-3,6-dichlorpyridazin
4-Amino-3,6-dichloropyridazine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163,9900 g/mol
MDL: MFCD01647253
CAS-Nummer: 823-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR