3-Brom-2-chlor-5-nitropyridin

Lieferant: COMBI-BLOCKS

OT-0072-5G OT-0072-25G OT-0072-1G OT-0072-100G
COBBOT-0072-5GEA 38 EUR
COBBOT-0072-5G COBBOT-0072-25G COBBOT-0072-1G COBBOT-0072-100G
3-Brom-2-chlor-5-nitropyridin
3-Bromo-2-chloro-5-nitropyridine
Formel: C₅H₂BrClN₂O₂
Molecular Weight: 237.44 g/mol
MDL: MFCD00233989
CAS-Nummer: 5470-17-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR