4-Brom-N-tert-butylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
OT-0086-5G
OT-0086-1G
COBBOT-0086-5GEA
385
EUR
COBBOT-0086-5G
COBBOT-0086-1G
4-Brom-N-tert-butylbenzolsulfonamid
4-Brom-N-tert-butylbenzolsulfonamid
Formel:
C₁₀H₁₄BrNO₂S |
MDL:
MFCD00458267 CAS-Nummer: 93281-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...