2-(4-Bromphenylsulfonamido)essigsäure
Lieferant: COMBI-BLOCKS
OT-0095-5G
OT-0095-25G
OT-0095-1G
COBBOT-0095-5GEA
102
EUR
COBBOT-0095-5G
COBBOT-0095-25G
COBBOT-0095-1G
2-(4-Bromphenylsulfonamido)essigsäure
2-(4-Bromphenylsulfonamido)essigsäure
Formel:
C₈H₈BrNO₄S |
MDL:
MFCD01935943 CAS-Nummer: 13029-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...