2-(4-Bromphenylsulfonamido)essigsäure

Lieferant: COMBI-BLOCKS

OT-0095-5G OT-0095-25G OT-0095-1G
COBBOT-0095-5GEA 102 EUR
COBBOT-0095-5G COBBOT-0095-25G COBBOT-0095-1G
2-(4-Bromphenylsulfonamido)essigsäure
2-(4-Bromphenylsulfonamido)essigsäure
Formel: C₈H₈BrNO₄S
MDL: MFCD01935943
CAS-Nummer: 13029-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR