3-Brom-N-cyclohexyl-4-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0161-5G OT-0161-1G
COBBOT-0161-5GEA 0 EUR
COBBOT-0161-5G COBBOT-0161-1G
3-Brom-N-cyclohexyl-4-methylbenzolsulfonamid
3-Brom-N-cyclohexyl-4-methylbenzolsulfonamid
Formel: C₁₃H₁₈BrNO₂S
MDL: MFCD07363834
CAS-Nummer: 850429-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR