2,4-Dibrom-6-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Amino-3,5-dibrom-α,α,α-trifluortoluol

OT-0184-25G
COBBOT-0184-25GEA 64.5 EUR
COBBOT-0184-25G
2,4-Dibrom-6-(trifluormethyl)anilin
2,4-Dibrom-6-(trifluormethyl)anilin
Formel: C₇H₄Br₂F₃N
Molecular Weight: 318,92 g/mol
MDL: MFCD00178769
CAS-Nummer: 71757-14-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR