3-Brom-5-chlorphenol

Lieferant: COMBI-BLOCKS

OT-0189-25G
COBBOT-0189-25GEA 198 EUR
COBBOT-0189-25G
3-Brom-5-chlorphenol
3-Brom-5-chlorphenol
Formel: C₆H₄BrClO
Molecular Weight: 207,45 g/mol
MDL: MFCD07780676
CAS-Nummer: 56962-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR