4-Brom-2-(morpholin-4-ylsulfonyl)anisol
Lieferant: COMBI-BLOCKS
Synonyme:
4-((5-Bromo-2-methoxyphenyl)sulphonyl)morpholine
OT-0197-5G
OT-0197-1G
COBBOT-0197-5GEA
692
EUR
COBBOT-0197-5G
COBBOT-0197-1G
4-Brom-2-(morpholin-4-ylsulfonyl)anisol
4-Brom-2-(morpholin-4-ylsulfonyl)anisol
Formel:
C₁₁H₁₄BrNO₄S Molecular Weight: 336.2000 g/mol |
MDL:
MFCD02043681 CAS-Nummer: 325809-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...