6-Amino-3-brom-2-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

OT-0319-100G OT-0319-25G OT-0319-5G
COBBOT-0319-100GEA 0 EUR
COBBOT-0319-100G COBBOT-0319-25G COBBOT-0319-5G
6-Amino-3-brom-2-methylpyridin
6-Amino-3-brom-2-methylpyridin
Formel: C₆H₇BrN₂
MDL: MFCD00068230
CAS-Nummer: 42753-71-9
EINECS: 255-927-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR