1,3-Dibrom-5-(trifluormethoxy)benzol
Lieferant: COMBI-BLOCKS
OT-0339-5G
OT-0339-25G
OT-0339-100G
COBBOT-0339-5GEA
26.2
EUR
COBBOT-0339-5G
COBBOT-0339-25G
COBBOT-0339-100G
1,3-Dibrom-5-(trifluormethoxy)benzol
1,3-Dibromo-5-(trifluoromethoxy)benzene
Formel:
C₇H₃Br₂F₃O Molecular Weight: 319.90 g/mol |
MDL:
MFCD00075280 CAS-Nummer: 207226-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...