5-Brom-2-hydroxypyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 3-Brom-6-pyridinol, 3-Brom-6-hydroxypyridin, 5-Brom-2-pyridinol

OT-0365-25G
COBBOT-0365-25GEA 38.2 EUR
COBBOT-0365-25G
5-Brom-2-hydroxypyridin
5-Brom-2-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
MDL: MFCD00234042
CAS-Nummer: 13466-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR