1-Brom-3-chlor-2-fluorbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-2-fluorobromobenzene

OT-0395-5G
COBBOT-0395-5GEA 0 EUR
COBBOT-0395-5G
1-Brom-3-chlor-2-fluorbenzol
1-Bromo-3-chloro-2-fluorobenzene
Formel: C₆H₃BrClF
Molecular Weight: 209,44 g/mol
MDL: MFCD03412182
CAS-Nummer: 144584-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR