2-(Methylsulfonyl)phenol

Lieferant: COMBI-BLOCKS

OT-0401-1G OT-0401-5G
COBBOT-0401-1GEA 0 EUR
COBBOT-0401-1G COBBOT-0401-5G
2-(Methylsulfonyl)phenol
Formel: C₇H₈O₃S
MDL: MFCD00127516
CAS-Nummer: 27489-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR