1-(Boc-amino)-4-bromnaphthalin

Lieferant: COMBI-BLOCKS

OT-0418-1G
COBBOT-0418-1GEA 623 EUR
COBBOT-0418-1G
1-(Boc-amino)-4-bromnaphthalin
1-(Boc-amino)-4-bromnaphthalin
Formel: C₁₅H₁₆BrNO₂
MDL: MFCD08704822
CAS-Nummer: 168169-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR