3-Brom-5-(trifluormethyl)benzaldehyd

Lieferant: COMBI-BLOCKS

OT-0424-5G OT-0424-25G OT-0424-1G
COBBOT-0424-5GEA 0 EUR
COBBOT-0424-5G COBBOT-0424-25G COBBOT-0424-1G
3-Brom-5-(trifluormethyl)benzaldehyd
3-Bromo-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₄BrF₃O
MDL: MFCD08059506
CAS-Nummer: 477535-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR