4-Brom-3-fluorthioanisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-2-fluor-4-(methylthio)benzol

OT-0448-1G
COBBOT-0448-1GEA 265 EUR
COBBOT-0448-1G
4-Brom-3-fluorthioanisol
4-Brom-3-fluorthioanisol
Formel: C₇H₆BrFS
MDL: MFCD09027283
CAS-Nummer: 917562-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR