2-Adamantyl-4-brom-1-(2,2,2-trifluoroethoxy)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 1-[5-Bromo-2-(2,2,2-trifluoroethoxy)phenyl]adamantane

OT-0467-1G
COBBOT-0467-1GEA 623 EUR
COBBOT-0467-1G
2-Adamantyl-4-brom-1-(2,2,2-trifluoroethoxy)benzol
2-Adamantyl-4-brom-1-(2,2,2-trifluoroethoxy)benzol
Formel: C₁₈H₂₀BrF₃O
MDL: MFCD09258647
CAS-Nummer: 929000-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR