2-(tert-Butyldimethylsilyloxy)-6-bromnaphthalin
Lieferant: COMBI-BLOCKS
OT-0474-5G
OT-0474-1G
OT-0474-25G
COBBOT-0474-5GEA
74.2
EUR
COBBOT-0474-5G
COBBOT-0474-1G
COBBOT-0474-25G
2-(tert-Butyldimethylsilyloxy)-6-bromnaphthalin
2-(tert-Butyldimethylsilyloxy)-6-bromnaphthalin
Formel:
C₁₆H₂₁BrOSi |
MDL:
MFCD11044858 CAS-Nummer: 100751-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...