4-Brom-3-methylbenzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-2-methyl-4-(trifluoromethyl)benzene

OT-0506-1G
COBBOT-0506-1GEA 38.5 EUR
COBBOT-0506-1G
4-Brom-3-methylbenzotrifluorid
4-Brom-3-methylbenzotrifluorid
Formel: C₈H₆BrF₃
Molecular Weight: 239,0325 g/mol
MDL: MFCD09258773
CAS-Nummer: 929000-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR