1-Brom-2-methyl-4-(trifluormethoxy)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-5-(trifluoromethoxy)toluene

OT-0529-5G OT-0529-1G
COBBOT-0529-5GEA 149 EUR
COBBOT-0529-5G COBBOT-0529-1G
1-Brom-2-methyl-4-(trifluormethoxy)benzol
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene
Formel: C₈H₆BrF₃O
Molecular Weight: 255.0319 g/mol
MDL: MFCD01631545
CAS-Nummer: 261951-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR