1,4-Dibrom-2,5-bis(brommethyl)benzol

Lieferant: COMBI-BLOCKS

OT-0536-5G OT-0536-1G
COBBOT-0536-5GEA 587 EUR
COBBOT-0536-5G COBBOT-0536-1G
1,4-Dibrom-2,5-bis(brommethyl)benzol
1,4-Dibrom-2,5-bis(brommethyl)benzol
Formel: C₈H₆Br₄
Molecular Weight: 421.7500 g/mol
MDL: MFCD03411745
CAS-Nummer: 35335-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR