3-Chlor-2-fluorthioanisol
Lieferant: COMBI-BLOCKS
OT-0539-5G
OT-0539-1G
COBBOT-0539-5GEA
937
EUR
COBBOT-0539-5G
COBBOT-0539-1G
3-Chlor-2-fluorthioanisol
3-Chlor-2-fluorthioanisol
Formel:
C₇H₆ClFS Molecular Weight: 176.64 g/mol |
MDL:
MFCD09475846 CAS-Nummer: 214057-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...